5-Bromo-2H-1,3-benzodioxol-4-ol ≥97%

Supplier: AOBChem USA

Warning

26948-1G 26948-25G 26948-500MG 26948-5G
76929-352EA 442.4 USD
76929-352 76929-354 76929-356 76929-358
5-Bromo-2H-1,3-benzodioxol-4-ol ≥97%
5-Bromo-2H-1,3-benzodioxol-4-ol

Precaución: for Research Use Only

Formula: C₇H₅BrO₃
MW: 217.017 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24714309
CAS Number: 82299-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR