4-Bromo-2-formylthiazole ≥97%

Supplier: AOBChem USA

Warning

26891-25G 26891-5G
76929-342EA 304.48 USD
76929-342 76929-344
4-Bromo-2-formylthiazole ≥97%
4-Bromo-2-formylthiazole
Formula: C₄H₂BrNOS
Storage Temperature: Refrigerator
MDL Number: MFCD06660122
CAS Number: 167366-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR