4-Isopropoxy-3-methylphenol ≥95%

Supplier: AOBChem USA

Warning

26829-1G 26829-25G 26829-500MG 26829-5G
76929-298EA 155.96 USD
76929-298 76929-300 76929-302 76929-304
4-Isopropoxy-3-methylphenol ≥95%
4-Isopropoxy-3-methylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD14535469
CAS Number: 1216201-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR