4-Cyano-2-methylphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 4-Cyano-2-methylphenylboronic acid pinacol ester

26553-1G 26553-25G 26553-5G
76929-262EA 62.6 USD
76929-262 76929-264 76929-266
4-Cyano-2-methylphenylboronic acid, pinacol ester ≥97%
4-Cyano-2-methylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₂
MW: 243.109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18729899
CAS Number: 848953-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR