4-Fluoro-3-(phenylmethoxy)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

26533-1G 26533-25G 26533-500MG 26533-5G
76929-236EA 224.92 USD
76929-236 76929-238 76929-240 76929-242
4-Fluoro-3-(phenylmethoxy)benzenemethanol ≥95%
4-Fluoro-3-(phenylmethoxy)benzenemethanol

Precaución: for Research Use Only

Formula: C₁₄H₁₃FO₂
MW: 232.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27756664
CAS Number: 959251-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR