5-Bromo-2-iodophenol ≥97%

Supplier: AOBChem USA

Warning

26522-25G 26522-5G
76929-216EA 310.85 USD
76929-216 76929-218
5-Bromo-2-iodophenol ≥97%
5-Bromo-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
MW: 298.904 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16999931
CAS Number: 858855-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR