2-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-(2-Hydroxypropan-2-yl)pyridine-3-boronic acid pinacol ester

26447-1G 26447-25G 26447-500MG 26447-5G
76929-202EA 155.96 USD
76929-202 76929-204 76929-206 76929-208
2-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)propan-2-ol ≥95%
2-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-yl)propan-2-ol
Formula: C₁₄H₂₂BNO₃
MW: 263.14 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15143417
CAS Number: 1257431-63-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR