4-Chloronicotinaldehyde hydrochloride ≥97%

Supplier: AOBChem USA

Warning

26396-1G 26396-25G 26396-5G
76929-190EA 64.72 USD
76929-190 76929-192 76929-194
4-Chloronicotinaldehyde hydrochloride ≥97%
4-Chloronicotinaldehyde hydrochloride

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD23703143
CAS Number: 1449008-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR