4-Hydroxymethyl-3-ethylphenylboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
26284-1G
26284-25G
26284-500MG
26284-5G
76929-158EA
409.51
USD
76929-158
76929-160
76929-162
76929-164
4-Hydroxymethyl-3-ethylphenylboronic acid pinacol ester ≥97%
4-Hydroxymethyl-3-ethylphenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₃BO₃ Storage Temperature: Ambient |
MDL Number:
MFCD26045271 CAS Number: 2121511-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...