4-Chloro-3,5-difluorobenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
26239-25G
26239-5G
76929-132EA
244.01
USD
76929-132
76929-134
4-Chloro-3,5-difluorobenzoic acid ≥97%
4-Chloro-3,5-difluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃ClF₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD11554154 CAS Number: 1160573-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...