6-Bromo-2-trifluoromethyl-1H-benzoimidazole ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-(trifluoromethyl)-1H-benzimidazole

26237-1G 26237-25G 26237-5G
76929-122EA 81.69 USD
76929-122 76929-124 76929-126
6-Bromo-2-trifluoromethyl-1H-benzoimidazole ≥95% research grade
6-Bromo-2-trifluoromethyl-1H-benzoimidazole
Formula: C₈H₄BrF₃N₂
MW: 265.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00460102
CAS Number: 3671-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR