4-Bromo-2-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

26159-1G 26159-25G 26159-500MG 26159-5G
76929-082EA 155.96 USD
76929-082 76929-084 76929-086 76929-088
4-Bromo-2-isopropoxybenzaldehyde ≥97%
4-Bromo-2-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
MDL Number: MFCD11651784
CAS Number: 1099672-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR