4-Chloro-1H-indazol-7-ylboronic acid ≥97%

Supplier: AOBChem USA

Warning

26115-1G 26115-500MG 26115-5G
76929-064EA 676.86 USD
76929-064 76929-066 76929-068
4-Chloro-1H-indazol-7-ylboronic acid ≥97%
4-Chloro-1H-indazol-7-ylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BClN₂O₂
MW: 196.399 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26045253
CAS Number: 2121513-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR