5-Bromo-2-fluoro-4-methylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
26108-1G
26108-25G
26108-500MG
26108-5G
76929-054EA
153.84
USD
76929-054
76929-056
76929-058
76929-060
5-Bromo-2-fluoro-4-methylbenzoic acid ≥97%
5-Bromo-2-fluoro-4-methylbenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₆BrFO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD23712067 CAS Number: 515135-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...