5-Bromo-2-chloro-4-fluoroaniline ≥97%

Supplier: AOBChem USA

Warning

26079-1G 26079-25G 26079-500MG 26079-5G
76929-028EA 124.13 USD
76929-028 76929-030 76929-032 76929-034
5-Bromo-2-chloro-4-fluoroaniline ≥97%
5-Bromo-2-chloro-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrClFN
Storage Temperature: Refrigerator
MDL Number: MFCD21603962
CAS Number: 305795-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR