4-Bromo-2-fluoro-6-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

26046-1G 26046-25G 26046-500MG 26046-5G
76929-014EA 141.1 USD
76929-014 76929-016 76929-018 76929-020
4-Bromo-2-fluoro-6-methoxybenzonitrile ≥97%
4-Bromo-2-fluoro-6-methoxybenzonitrile
Formula: C₈H₅BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD16619400
CAS Number: 457051-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR