4-Chloro-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

25961-1G 25961-25G 25961-500MG 25961-5G
76929-002EA 290.69 USD
76929-002 76929-004 76929-006 76929-008
4-Chloro-3-methylbenzyl alcohol ≥97%
4-Chloro-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉ClO
Storage Temperature: Refrigerator
MDL Number: MFCD06201141
CAS Number: 131271-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR