5-Chloro-[1,1'-biphenyl]-2-amine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-2-phenylaniline

25879-1G 25879-25G 25879-500MG 25879-5G
76928-974EA 259.93 USD
76928-974 76928-976 76928-978 76928-980
5-Chloro-[1,1'-biphenyl]-2-amine ≥97% research grade
5-Chloro-[1,1'-biphenyl]-2-amine
Formula: C₁₂H₁₀ClN
MW: 203.67 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23713168
CAS Number: 73006-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR