5-Chloro-2,4-difluorophenol ≥97%

Supplier: AOBChem USA

Warning

25850-1G 25850-500MG 25850-5G
76928-944EA 647.15 USD
76928-944 76928-946 76928-948
5-Chloro-2,4-difluorophenol ≥97%
5-Chloro-2,4-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃ClF₂O
MW: 164.537 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18415632
CAS Number: 2268-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR