4-Bromo-3-chlorobenzyl Alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-Bromo-3-Chlorophenyl)methanol, 4-Bromo-3-chlorobenzyl alcohol

25839-25G 25839-5G
76928-936EA 375.56 USD
76928-936 76928-938
4-Bromo-3-chlorobenzyl Alcohol ≥97%
4-Bromo-3-chlorobenzyl Alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.479 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059111
CAS Number: 120077-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR