4-Bromo-2-isopropyl-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

25826-1G 25826-25G 25826-5G
76928-912EA 84.88 USD
76928-912 76928-914 76928-916
4-Bromo-2-isopropyl-1-methoxybenzene ≥97%
4-Bromo-2-isopropyl-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11846382
CAS Number: 24591-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR