5-Cyclopropylpyridine-3-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

25780-1G 25780-500MG 25780-5G
76928-886EA 498.63 USD
76928-886 76928-888 76928-890
5-Cyclopropylpyridine-3-carbaldehyde ≥95%
5-Cyclopropylpyridine-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.174 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18255592
CAS Number: 1211589-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR