4-Methyl-2-phenylaniline ≥97%

Supplier: AOBChem USA

Warning

25619-1G 25619-25G 25619-500MG 25619-5G
76928-842EA 231.28 USD
76928-842 76928-844 76928-846 76928-848
4-Methyl-2-phenylaniline ≥97%
4-Methyl-2-phenylaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₃N
MW: 183.249 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06408241
CAS Number: 42308-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR