5-Bromo-2-phenylpyridine ≥95%

Supplier: AOBChem USA

Warning

25472-25G 25472-5G
76928-786EA 605.78 USD
76928-786 76928-788
5-Bromo-2-phenylpyridine ≥95%
5-Bromo-2-phenylpyridine
Formula: C₁₁H₈BrN
MW: 234.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114181
CAS Number: 27012-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR