4-Bromo-3-fluoro-2-methylaniline ≥97%

Supplier: AOBChem USA

Warning

25406-25G 25406-5G
76928-778EA 381.93 USD
76928-778 76928-780
4-Bromo-3-fluoro-2-methylaniline ≥97%
4-Bromo-3-fluoro-2-methylaniline
Formula: C₇H₇BrFN
Storage Temperature: Ambient
MDL Number: MFCD09800761
CAS Number: 127408-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR