5-Bromo-2-fluoro-4-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

25368-1G 25368-25G 25368-500MG 25368-5G
76928-764EA 241.89 USD
76928-764 76928-766 76928-768 76928-770
5-Bromo-2-fluoro-4-hydroxybenzaldehyde ≥97%
5-Bromo-2-fluoro-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrFO₂
Storage Temperature: Ambient
CAS Number: 914397-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR