Methyl 4-bromo-3,5-dichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-3,5-dichloro-benzoic acid methyl ester

25303-1G 25303-25G 25303-5G
76928-738EA 70.02 USD
76928-738 76928-740 76928-742
Methyl 4-bromo-3,5-dichlorobenzoate ≥97%
Methyl 4-bromo-3,5-dichlorobenzoate
Formula: C₈H₅BrCl₂O₂
MW: 283.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14704248
CAS Number: 117738-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR