4-Bromo-3,5-dichloroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

25284-1G 25284-25G 25284-5G
76928-724EA 109.28 USD
76928-724 76928-726 76928-728
4-Bromo-3,5-dichloroiodobenzene ≥97%
4-Bromo-3,5-dichloroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂I
Storage Temperature: Refrigerator
CAS Number: 1000574-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR