4-Methoxy-2-(methoxymethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

25248-1G 25248-25G 25248-500MG 25248-5G
76928-700EA 260.99 USD
76928-700 76928-702 76928-704 76928-706
4-Methoxy-2-(methoxymethyl)phenylboronic acid ≥97%
4-Methoxy-2-(methoxymethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD21187812
CAS Number: 1451392-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR