4-Chloro-2-methylthio-1-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

25185-1G 25185-25G 25185-500MG 25185-5G
76928-684EA 178.24 USD
76928-684 76928-686 76928-688 76928-690
4-Chloro-2-methylthio-1-(trifluoromethyl)benzene ≥97%
4-Chloro-2-methylthio-1-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₆ClF₃S
MW: 226.646 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22488885
CAS Number: 1428234-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR