5-Cyano-2-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

25109-1G 25109-25G 25109-500MG 25109-5G
76928-652EA 198.39 USD
76928-652 76928-654 76928-656 76928-658
5-Cyano-2-methylbenzoic acid ≥95%
5-Cyano-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189533
CAS Number: 1975-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR