4-Bromo-2-ethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

24934-1G 24934-25G 24934-5G
76928-596EA 62.6 USD
76928-596 76928-598 76928-600
4-Bromo-2-ethylbenzoic acid ≥97%
4-Bromo-2-ethylbenzoic acid
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18410376
CAS Number: 644984-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR