2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Cyano-3-methylphenylboronic acid pinacol ester

24915-1G 24915-25G 24915-500MG 24915-5G
76928-586EA 144.29 USD
76928-586 76928-588 76928-590 76928-592
2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile ≥97%
2-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Formula: C₁₄H₁₈BNO₂
MW: 243.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11035951
CAS Number: 775351-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR