5-Cyclopropyl-2-fluoropyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

24896-1G 24896-25G 24896-500MG 24896-5G
76928-570EA 263.11 USD
76928-570 76928-572 76928-574 76928-576
5-Cyclopropyl-2-fluoropyridine-3-boronic acid ≥97%
5-Cyclopropyl-2-fluoropyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BFNO₂
MW: 180.972 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24448899
CAS Number: 2121511-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR