5-Bromo-2-fluoro-3-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

24882-1G 24882-25G 24882-500MG 24882-5G
76928-558EA 350.1 USD
76928-558 76928-560 76928-562 76928-564
5-Bromo-2-fluoro-3-phenylpyridine ≥97%
5-Bromo-2-fluoro-3-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₇BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD18258861
CAS Number: 473596-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR