4-Chloro-3-isopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24865-1G 24865-25G 24865-500MG 24865-5G
76928-542EA 409.51 USD
76928-542 76928-544 76928-546 76928-548
4-Chloro-3-isopropylbenzaldehyde ≥97%
4-Chloro-3-isopropylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO
MW: 182.647 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19686193
CAS Number: 1289158-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR