5-Bromo-2-fluoro-3-isopropoxypyridine ≥95%

Supplier: AOBChem USA

Warning

24707-1G 24707-25G 24707-500MG 24707-5G
76928-486EA 159.14 USD
76928-486 76928-488 76928-490 76928-492
5-Bromo-2-fluoro-3-isopropoxypyridine ≥95%
5-Bromo-2-fluoro-3-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₉BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD22543741
CAS Number: 1394291-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR