5-Bromo-4-methyl-2-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

24566-1G 24566-25G 24566-500MG 24566-5G
76928-446EA 224.92 USD
76928-446 76928-448 76928-450 76928-452
5-Bromo-4-methyl-2-(methylthio)pyridine ≥95%
5-Bromo-4-methyl-2-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNS
MW: 218.114 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543729
CAS Number: 247135-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR