4-Bromo-5-chloro-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

24502-1G 24502-25G 24502-500MG 24502-5G
76928-398EA 350.1 USD
76928-398 76928-400 76928-402 76928-404
4-Bromo-5-chloro-2-methylphenylboronic acid ≥97%
4-Bromo-5-chloro-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrClO₂
MW: 249.297 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22543713
CAS Number: 1451391-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR