5-Chloro-2-(trifluoromethyl)benzamide ≥97% research grade

Supplier: AOBChem USA

Warning

24449-25G 24449-5G
76928-386EA 418 USD
76928-386 76928-388
5-Chloro-2-(trifluoromethyl)benzamide ≥97% research grade
5-Chloro-2-(trifluoromethyl)benzamide
Formula: C₈H₅ClF₃NO
Storage Temperature: Ambient
MDL Number: MFCD01631406
CAS Number: 654-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR