5-Bromo-4-chloro-2-iodotoluene ≥97%

Supplier: AOBChem USA

Warning

24404-1G 24404-25G 24404-5G
76928-376EA 66.84 USD
76928-376 76928-378 76928-380
5-Bromo-4-chloro-2-iodotoluene ≥97%
5-Bromo-4-chloro-2-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrClI
Storage Temperature: Ambient
MDL Number: MFCD09800822
CAS Number: 1000578-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR