4-Chloro-3-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

24365-1G 24365-25G 24365-500MG 24365-5G
76928-362EA 198.39 USD
76928-362 76928-364 76928-366 76928-368
4-Chloro-3-ethoxybenzaldehyde ≥97%
4-Chloro-3-ethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16619399
CAS Number: 85259-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR