(5-Bromo-4-methylpyridin-2-yl)methanol ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
5-Bromo-2-hydroxymethyl-4-methylpyridine
24324-1G
24324-25G
24324-500MG
24324-5G
76928-346EA
135.8
USD
76928-346
76928-348
76928-350
76928-352
(5-Bromo-4-methylpyridin-2-yl)methanol ≥97%
(5-Bromo-4-methylpyridin-2-yl)methanol
Formula:
C₇H₈BrNO MW: 202.04 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22543690 CAS Number: 1394291-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...