5-Bromo-4-methylpicolinaldehyde ≥97%

Supplier: AOBChem USA

Warning

24321-1G 24321-25G 24321-5G
76928-334EA 109.28 USD
76928-334 76928-336 76928-338
5-Bromo-4-methylpicolinaldehyde ≥97%
5-Bromo-4-methylpicolinaldehyde

Precaución: for Research Use Only

Formula: C₇H₆BrNO
Storage Temperature: Refrigerator
MDL Number: MFCD07375095
CAS Number: 886364-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR