5-Bromo-3-fluoro-2-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-1-fluoro-2-iodo-3-methoxybenzene

24263-1G 24263-25G 24263-5G
76928-324EA 64.72 USD
76928-324 76928-326 76928-328
5-Bromo-3-fluoro-2-iodoanisole ≥97%
5-Bromo-3-fluoro-2-iodoanisole

Precaución: for Research Use Only

Formula: C₇H₅BrFIO
Storage Temperature: Refrigerator
MDL Number: MFCD22543687
CAS Number: 1394291-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR