5-Bromo-2-(2-methoxyethoxy)pyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
24210-1G
24210-25G
24210-5G
76928-310EA
85.94
USD
76928-310
76928-312
76928-314
5-Bromo-2-(2-methoxyethoxy)pyridine ≥97%
5-Bromo-2-(2-methoxyethoxy)pyridine
Formula:
C₈H₁₀BrNO₂ MW: 232.07 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13190750 CAS Number: 212961-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...