Methyl 4-phenoxybenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

24187-1G 24187-25G 24187-5G
76928-302EA 88.06 USD
76928-302 76928-304 76928-306
Methyl 4-phenoxybenzoate ≥97% research grade
Methyl 4-phenoxybenzoate
Formula: C₁₄H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00446993
CAS Number: 21218-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR