3-Bromo-5-(methylsulfonyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-3-methylsulphonylpyridine, 5-Bromopyridin-3-yl methyl sulphone

24044-1G 24044-25G 24044-5G
76928-288EA 117.76 USD
76928-288 76928-290 76928-292
3-Bromo-5-(methylsulfonyl)pyridine ≥97%
3-Bromo-5-(methylsulfonyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MW: 236.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09801057
CAS Number: 445491-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR