5-(Methylthio)thiophene-2-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

23970-25G 23970-5G
76928-270EA 595.17 USD
76928-270 76928-272
5-(Methylthio)thiophene-2-carboxaldehyde ≥97%
5-(Methylthio)thiophene-2-carboxaldehyde
Formula: C₆H₆OS₂
Storage Temperature: Ambient
MDL Number: MFCD00173744
CAS Number: 24445-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR