5-Fluoro-2-iodoanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-1-iodo-2-methoxybenzene

23963-25G 23963-5G
76928-264EA 569.71 USD
76928-264 76928-266
5-Fluoro-2-iodoanisole ≥97%
5-Fluoro-2-iodoanisole
Formula: C₇H₆FIO
MW: 252.025 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18396916
CAS Number: 450-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR